Gene Transcripts
- Gene Symbol
- MIR670HG
- Entrez Gene ID
- 100507261
- Gene Name
- MIR670 host gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 43569306 - 43570400 on Build GRCh38
- UniGene ID
- Hs.664953
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_131246.1 |
|
2 |
684 |
|
GenBank mRNA |
BC031305.2 |
- |
2 |
684 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000533531.1 |
- |
2 |
684 |
|
Chr. 11 - 43569306 - 43570395 [+] |
View Full Product Details