Gene Transcripts

Gene Symbol
Spata20
Entrez Gene ID
217116
Gene Name
spermatogenesis associated 20
Gene Aliases
Tisp78
Chromosome Location
Chr. 11: 94478904 - 94485310 on Build GRCm38
UniGene ID
Mm.156123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144827.4 NP_659076.2 2 131
GenBank mRNA BC050788.1 - 2 131

View Full Product Details