Gene Transcripts

Gene Symbol
Sec16a
Entrez Gene ID
227648
Gene Name
SEC16 homolog A (S. cerevisiae)
Gene Aliases
AU024582, C230052J16Rik
Chromosome Location
Chr. 2: 26409431 - 26446162 on Build GRCm38
UniGene ID
Mm.206371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153125.2 NP_694765.2 2 3420
GenBank mRNA AK082458.1 - 1 46
BC059898.1 - 1 638
CO432077.1 - 1 192

View Full Product Details