Gene Transcripts

Gene Symbol
Zfp334
Entrez Gene ID
228876
Gene Name
zinc finger protein 334
Gene Aliases
D2Ertd535e
Chromosome Location
Chr. 2: 165377434 - 165388317 on Build GRCm38
UniGene ID
Mm.26316
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178411.3 NP_848498.2 5 1153
GenBank mRNA BC027792.1 - 1 488
BC040388.1 - 5 1153 3501307

View Full Product Details