Gene Transcripts
- Gene Symbol
- Olfr1034
- Entrez Gene ID
- 258216
- Gene Name
- olfactory receptor 1034
- Gene Aliases
- MOR227-8P, MOR245-14P, Olfr1533-ps1
- Chromosome Location
- Chr. 2: 86046444 - 86047466 on Build GRCm38
- UniGene ID
- Mm.330363
- Species
- Mus musculus
- Gene Symbol
- Olfr1036
- Entrez Gene ID
- 258245
- Gene Name
- olfactory receptor 1036
- Gene Aliases
- GA_x5J8B7TTB99-482-964, MOR245-25, MOR262-13
- Chromosome Location
- Chr. 2: 86074704 - 86075718 on Build GRCm38
- UniGene ID
- Mm.377389
- Species
- Mus musculus
View Full Product Details