Gene Transcripts

Gene Symbol
Olfr460
Entrez Gene ID
258381
Gene Name
olfactory receptor 460
Gene Aliases
MOR120-2
Chromosome Location
Chr. 6: 40569152 - 40572504 on Build GRCm38
UniGene ID
Mm.377469
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146383.1 NP_666495.1 1 878
GenBank mRNA BC119048.1 - 1 909
BC120738.1 - 1 909 40129661

View Full Product Details