Gene Transcripts
- Gene Symbol
- Olfr859
- Entrez Gene ID
- 258519
- Gene Name
- olfactory receptor 859
- Gene Aliases
- MOR146-3
- Chromosome Location
- Chr. 9: 19808320 - 19809249 on Build GRCm38
- UniGene ID
- Mm.491770
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146526.2 |
NP_666737.2
|
1 |
501 |
|
- Gene Symbol
- Olfr58
- Entrez Gene ID
- 18358
- Gene Name
- olfactory receptor 58
- Gene Aliases
- IG6, MOR146-3, MOR146-7P
- Chromosome Location
- Chr. 9: 19780283 - 19784064 on Build GRCm38
- UniGene ID
- Mm.248950
- Species
- Mus musculus
View Full Product Details