Gene Transcripts

Gene Symbol
Olfr1030
Entrez Gene ID
258581
Gene Name
olfactory receptor 1030
Gene Aliases
MOR196-2
Chromosome Location
Chr. 2: 85979312 - 85984798 on Build GRCm38
UniGene ID
Mm.223108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146588.2 NP_666799.1 2 280
GenBank mRNA BC128001.1 - 1 251

View Full Product Details