Gene Transcripts

Gene Symbol
Olfr508
Entrez Gene ID
258769
Gene Name
olfactory receptor 508
Gene Aliases
MOR204-6
Chromosome Location
Chr. 7: 108629994 - 108630926 on Build GRCm38
UniGene ID
Mm.223200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146773.1 NP_666984.1 1 823
GenBank mRNA BC130241.1 - 1 827

View Full Product Details