Gene Transcripts

Gene Symbol
Olfr472
Entrez Gene ID
258770
Gene Name
olfactory receptor 472
Gene Aliases
MOR204-5
Chromosome Location
Chr. 7: 107901558 - 107903703 on Build GRCm38
UniGene ID
Mm.389960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146774.1 NP_666985.1 1 131
GenBank mRNA BC138079.1 - 1 223
BC145756.1 - 1 223 40131059

View Full Product Details