Gene Transcripts

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
Chromosome Location
Chr. 3: 148697871 - 148743003 on Build GRCh38
UniGene ID
Hs.477887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000685.4 NP_000676.1 3 685
NM_004835.4 NP_004826.5 3 659
NM_009585.3 NP_033611.1 2 601
NM_031850.3 NP_114038.4 4 743
NM_032049.3 NP_114438.2 3 559
GenBank mRNA AK291541.1 - 3 654
BC022447.1 - 2 569 4799755
BC068494.1 - 3 654 30337806
D13814.1 - 1 536
M87290.1 - 2 577
M93394.1 - 3 469
S77410.1 - 3 631
X65699.1 - 2 534

View Full Product Details