Gene Transcripts

Gene Symbol
Gm5907
Entrez Gene ID
546036
Gene Name
predicted gene 5907
Gene Aliases
EG546036, Gm845
Chromosome Location
Chr. 8: 13997112 - 14003561 on Build GRCm38
UniGene ID
Mm.31785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK145481.1 - 3 607
BX520893.1 - 460

View Full Product Details