Gene Transcripts
- Gene Symbol
- Tpk1
- Entrez Gene ID
- 29807
- Gene Name
- thiamine pyrophosphokinase
- Gene Aliases
- Chromosome Location
- Chr. 6: 43344999 - 43666484 on Build GRCm38
- UniGene ID
- Mm.320979
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033192.1 |
- |
1 |
193 |
|
- Gene Symbol
- 4930571B16Rik
- Entrez Gene ID
- 613117
- Gene Name
- RIKEN cDNA 4930571B16 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.488109
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016268.1 |
- |
1 |
954 |
|
- Gene Symbol
- A230004M16Rik
- Entrez Gene ID
- 320922
- Gene Name
- RIKEN cDNA A230004M16 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.390351
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK049150.1 |
- |
1 |
193 |
|
- Gene Symbol
- A730099G02Rik
- Entrez Gene ID
- 319306
- Gene Name
- RIKEN cDNA A730099G02 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043485.1 |
- |
1 |
570 |
|
AK133136.1 |
- |
1 |
971 |
|
- Gene Symbol
- Gm7968
- Entrez Gene ID
- 666183
- Gene Name
- predicted gene 7968
- Gene Aliases
- EG666183
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036791.1 |
- |
1 |
660 |
|
- Gene Symbol
- Ncoa7
- Entrez Gene ID
- 211329
- Gene Name
- nuclear receptor coactivator 7
- Gene Aliases
- 9030406N13Rik
- Chromosome Location
- Chr. 10: 30645582 - 30805977 on Build GRCm38
- UniGene ID
- Mm.297903
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042791.1 |
- |
|
1669 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK029879.1 |
- |
|
929 |
|
Chr. - - [-] |
View Full Product Details