Gene Transcripts
- Gene Symbol
- Gm7609
- Entrez Gene ID
- 665378
- Gene Name
- predicted pseudogene 7609
- Gene Aliases
- EG665378
- Chromosome Location
- Chr. 1: 85199592 - 85215395 on Build GRCm38
- UniGene ID
- Mm.440912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK156902.1 |
- |
2 |
922 |
|
View Full Product Details