Gene Transcripts

Gene Symbol
Ak1
Entrez Gene ID
24183
Gene Name
adenylate kinase 1
Gene Aliases
Ak 1
Chromosome Location
Chr. 3: 11652143 - 11659135 on Build Rnor_6.0
UniGene ID
Rn.219384
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024349.3 NP_077325.2 271
GenBank mRNA BC089797.1 - 271
D13376.1 - 21

View Full Product Details