Gene Transcripts

Gene Symbol
Pcsk2
Entrez Gene ID
25121
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
Chromosome Location
Chr. 3: 137618891 - 137923385 on Build Rnor_6.0
UniGene ID
Rn.89052
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012746.1 NP_036878.1 4, 5 825
GenBank mRNA M76706.1 - 4, 5 825
M83746.1 - 4, 5 797

View Full Product Details