Gene Transcripts

Gene Symbol
Jak3
Entrez Gene ID
25326
Gene Name
Janus kinase 3
Gene Aliases
RATJAK3
Chromosome Location
Chr. 16: 20107468 - 20120678 on Build Rnor_6.0
UniGene ID
Rn.11159
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012855.2 NP_036987.2 2 183
XM_008771064.1 XP_008769286.1 2 1281
GenBank mRNA D28508.1 - 2 183

View Full Product Details