Gene Transcripts
- Gene Symbol
- Bmp3
- Entrez Gene ID
- 25667
- Gene Name
- bone morphogenetic protein 3
- Gene Aliases
- PBMP3
- Chromosome Location
- Chr. 14: 12362912 - 12387572 on Build Rnor_6.0
- UniGene ID
- Rn.208979
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017105.1 |
NP_058801.1
|
2 |
1038 |
|
GenBank mRNA |
D63860.1 |
- |
2 |
1038 |
|
View Full Product Details