Gene Transcripts
- Gene Symbol
- Lrp2
- Entrez Gene ID
- 29216
- Gene Name
- low density lipoprotein receptor-related protein 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 55665153 - 55822484 on Build Rnor_6.0
- UniGene ID
- Rn.26430
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030827.1 |
NP_110454.1
|
20 |
2991 |
|
GenBank mRNA |
L34049.1 |
- |
20 |
2991 |
|
View Full Product Details