Gene Transcripts
- Gene Symbol
- Snca
- Entrez Gene ID
- 29219
- Gene Name
- synuclein, alpha (non A4 component of amyloid precursor)
- Gene Aliases
- Chromosome Location
- Chr. 4: 90782412 - 90882539 on Build Rnor_6.0
- UniGene ID
- Rn.1827
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019169.2 |
NP_062042.1
|
2, 3 |
284 |
|
XM_006236591.2 |
XP_006236653.1
|
2, 3 |
565 |
|
GenBank mRNA |
AF007758.1 |
- |
1, 2 |
147 |
|
AY550005.1 |
- |
1, 2 |
147 |
|
AY550006.1 |
- |
1, 2 |
147 |
|
BC087682.1 |
- |
2, 3 |
284 |
7321273 |
FQ212580.1 |
- |
2, 3 |
261 |
|
FQ227096.1 |
- |
2, 3 |
275 |
|
S73007.1 |
- |
2, 3 |
215 |
|
S73008.1 |
- |
2, 3 |
215 |
|
View Full Product Details