Gene Transcripts

Gene Symbol
Pde3a
Entrez Gene ID
50678
Gene Name
phosphodiesterase 3A, cGMP inhibited
Gene Aliases
RNPDE3A
Chromosome Location
Chr. 4: 175431904 - 175703844 on Build Rnor_6.0
UniGene ID
Rn.44403
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017337.1 NP_059033.1 11 2335
XM_008763422.1 XP_008761644.1 11 2403
GenBank mRNA U38179.1 - 11 2335

View Full Product Details