Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
65026
Gene Name
RWD domain containing 3
Gene Aliases
Rsume, X2cr1
Chromosome Location
Chr. 2: 224710351 - 224723129 on Build Rnor_6.0
UniGene ID
Rn.54470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128152.1 NP_001121624.1 2 521
XM_008761472.1 XP_008759694.1 2 550
GenBank mRNA AF271158.1 - 1 97

View Full Product Details