Gene Transcripts

Gene Symbol
Gng5
Entrez Gene ID
79218
Gene Name
guanine nucleotide binding protein (G protein), gamma 5
Gene Aliases
Chromosome Location
Chr. 2: 252359976 - 252368473 on Build Rnor_6.0
UniGene ID
Rn.2695
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024377.1 NP_077353.1 3 505
GenBank mRNA DQ120493.1 - 3 450
DQ120494.1 - 3 450
M95780.1 - 3 505

View Full Product Details