Gene Transcripts

Gene Symbol
Ssr3
Entrez Gene ID
81784
Gene Name
signal sequence receptor, gamma
Gene Aliases
Chromosome Location
Chr. 2: 156010997 - 156023853 on Build Rnor_6.0
UniGene ID
Rn.3264
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031120.2 NP_112382.2 236
GenBank mRNA AY724496.1 - 244
BC062015.1 - 236 5621495
Z14030.1 - 244

View Full Product Details