Gene Transcripts

Gene Symbol
Bmp1
Entrez Gene ID
83470
Gene Name
bone morphogenetic protein 1
Gene Aliases
Chromosome Location
Chr. 15: 52166401 - 52210780 on Build Rnor_6.0
UniGene ID
Rn.9305
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031323.1 NP_112613.1 4 660
XM_006252285.1 XP_006252347.1 1 96
XM_008770834.1 XP_008769056.1 4 694

View Full Product Details