Gene Transcripts

Gene Symbol
Defb1
Entrez Gene ID
83687
Gene Name
defensin beta 1
Gene Aliases
Chromosome Location
Chr. 16: 75294385 - 75309127 on Build Rnor_6.0
UniGene ID
Rn.31800
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031810.1 NP_113998.1 2 174
GenBank mRNA AF068860.1 - 2 150
AF093536.1 - 2 174

View Full Product Details