Gene Transcripts

Gene Symbol
Slc17a6
Entrez Gene ID
84487
Gene Name
solute carrier family 17 (vesicular glutamate transporter), member 6
Gene Aliases
Dnpi, Vglut2
Chromosome Location
Chr. 1: 106998704 - 107038717 on Build Rnor_6.0
UniGene ID
Rn.19372
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053427.1 NP_445879.1 7 1285
XM_006229265.1 XP_006229327.1 7 814
GenBank mRNA AF271235.1 - 7 1285

View Full Product Details