Gene Transcripts

Gene Symbol
Cdc42bpb
Entrez Gene ID
113960
Gene Name
CDC42 binding protein kinase beta (DMPK-like)
Gene Aliases
Chromosome Location
Chr. 6: 135746743 - 135830143 on Build Rnor_6.0
UniGene ID
Rn.105815
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053620.3 NP_446072.2 12 1607
XM_006240562.2 XP_006240624.1 12 1756
GenBank mRNA AF021936.1 - 12 1567
AY277590.1 - 12 1607

View Full Product Details