Gene Transcripts

Gene Symbol
Il5ra
Entrez Gene ID
114103
Gene Name
interleukin 5 receptor, alpha
Gene Aliases
Chromosome Location
Chr. 4: 138798769 - 138830636 on Build Rnor_6.0
UniGene ID
Rn.67068
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053645.1 NP_446097.1 2 97
XM_006236977.2 XP_006237039.1 4 488
GenBank mRNA AB056101.1 - 2 97
AF324153.1 - 2 97

View Full Product Details