Gene Transcripts

Gene Symbol
Kcnj9
Entrez Gene ID
116560
Gene Name
potassium channel, inwardly rectifying subfamily J, member 9
Gene Aliases
Girk3, Kir3.3
Chromosome Location
Chr. 13: 90701432 - 90710148 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053834.1 NP_446286.1 3 555
XM_008769730.1 XP_008767952.1 1 1517
GenBank mRNA L77929.1 - 3 555

View Full Product Details