Gene Transcripts

Gene Symbol
Lcn2
Entrez Gene ID
170496
Gene Name
lipocalin 2
Gene Aliases
Sip24
Chromosome Location
Chr. 3: 11414189 - 11417534 on Build Rnor_6.0
UniGene ID
Rn.11303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130741.1 NP_570097.1 1 117
GenBank mRNA BC089053.1 - 1 179
X13295.1 - 1 117

View Full Product Details