Gene Transcripts
- Gene Symbol
- Pcsk4
- Entrez Gene ID
- 171085
- Gene Name
- proprotein convertase subtilisin/kexin type 4
- Gene Aliases
- Chromosome Location
- Chr. 7: 12247312 - 12255729 on Build Rnor_6.0
- UniGene ID
- Rn.2899
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133559.1 |
NP_598243.1
|
13, 14 |
1701 |
|
XM_006240839.2 |
XP_006240901.1
|
13, 14 |
1917 |
|
XM_006240843.2 |
XP_006240905.1
|
13, 14 |
1887 |
|
XM_008765083.1 |
XP_008763305.1
|
12, 13 |
1989 |
|
XM_008765085.1 |
XP_008763307.1
|
12, 13 |
1959 |
|
XM_008765086.1 |
XP_008763308.1
|
12, 13 |
1956 |
|
XM_008765087.1 |
XP_008763309.1
|
12, 13 |
1989 |
|
XM_008765088.1 |
XP_008763310.1
|
12, 13 |
1989 |
|
GenBank mRNA |
BC097288.1 |
- |
13, 14 |
1713 |
|
L14937.1 |
- |
13, 14 |
1701 |
|
View Full Product Details