Gene Transcripts

Gene Symbol
Gchfr
Entrez Gene ID
171128
Gene Name
GTP cyclohydrolase I feedback regulator
Gene Aliases
Chromosome Location
Chr. 3: 110975923 - 110979957 on Build Rnor_6.0
UniGene ID
Rn.11137
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133595.2 NP_598279.1 3 195
GenBank mRNA BC086944.1 - 3 176
U53710.1 - 3 195
U85512.1 - 3 222

View Full Product Details