Gene Transcripts
- Gene Symbol
- Gpc2
- Entrez Gene ID
- 171517
- Gene Name
- glypican 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 19593041 - 19599374 on Build Rnor_6.0
- UniGene ID
- Rn.88131
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138511.1 |
NP_612520.1
|
7 |
1382 |
|
GenBank mRNA |
L20468.1 |
- |
7 |
1382 |
|
View Full Product Details