Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
171562
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 15: 19619947 - 19655381 on Build Rnor_6.0
UniGene ID
Rn.64648
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138528.1 NP_612537.1 14 1144
GenBank mRNA AF489855.1 - 15 1131
AY071924.1 - 14 1144
AY071925.1 - 11 826
FQ228432.1 - 989

View Full Product Details