Gene Transcripts

Gene Symbol
Sorbs3
Entrez Gene ID
282843
Gene Name
sorbin and SH3 domain containing 3
Gene Aliases
Sh3d4
Chromosome Location
Chr. 15: 51859004 - 51892653 on Build Rnor_6.0
UniGene ID
Rn.63481
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005762.1 NP_001005762.1 17 1561
XM_006252241.2 XP_006252303.1 18 3245
XM_006252242.1 XP_006252304.1 17 1652
XM_006252244.2 XP_006252306.1 16 1880
XM_006252246.1 XP_006252308.1 17 1597
XM_006252248.1 XP_006252310.1 5 445
XM_008770819.1 XP_008769041.1 15 3354
GenBank mRNA AF439378.1 - 16 1377
BC083666.1 - 17 1561 7135547

View Full Product Details