Gene Transcripts
- Gene Symbol
- TSPAN6
- Entrez Gene ID
- 7105
- Gene Name
- tetraspanin 6
- Gene Aliases
- T245, TM4SF6, TSPAN-6
- Chromosome Location
- Chr. X: 100627108 - 100637104 on Build GRCh38
- UniGene ID
- Hs.43233
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278740.1 |
NP_001265669.1
|
6, 7 |
1129 |
|
NM_001278741.1 |
NP_001265670.1
|
6, 7 |
899 |
|
NM_003270.3 |
NP_003261.1
|
6, 7 |
837 |
|
XM_011531018.1 |
XP_011529320.1
|
7, 8 |
639 |
|
GenBank mRNA |
AB097016.1 |
- |
6, 7 |
733 |
|
AB209876.1 |
- |
6, 7 |
796 |
|
AF043906.1 |
- |
6, 7 |
741 |
|
AF053453.1 |
- |
6, 7 |
693 |
|
AF133426.1 |
- |
6, 7 |
700 |
|
AK075072.1 |
- |
6, 7 |
786 |
|
AK299176.1 |
- |
7, 8 |
556 |
|
AK300852.1 |
- |
7, 8 |
777 |
|
AK309581.1 |
- |
6, 7 |
1112 |
|
AK315773.1 |
- |
6, 7 |
819 |
|
AY358825.1 |
- |
6, 7 |
714 |
|
BC012389.1 |
- |
6, 7 |
777 |
3857537 |
BT006977.1 |
- |
6, 7 |
674 |
|
CR457115.1 |
- |
6, 7 |
674 |
|
CX868833.1 |
- |
6, 7 |
608 |
|
U84895.1 |
- |
6, 7 |
741 |
|
View Full Product Details