Gene Transcripts

Gene Symbol
Cramp1l
Entrez Gene ID
287127
Gene Name
Crm, cramped-like (Drosophila)
Gene Aliases
Chromosome Location
Chr. 10: 14324799 - 14373411 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246051.2 XP_006246113.1 10 2514
XM_006246052.2 XP_006246114.1 10 2178
XM_006246053.2 XP_006246115.1 10 1964
XM_008767606.1 XP_008765828.1 10 2514

View Full Product Details