Gene Transcripts

Gene Symbol
Rnasek
Entrez Gene ID
287453
Gene Name
ribonuclease, RNase K
Gene Aliases
RGD1305687
Chromosome Location
Chr. 10: 56848367 - 56850085 on Build Rnor_6.0
UniGene ID
Rn.1025
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137561.2 NP_001131033.2 3 289
GenBank mRNA BC169131.1 - 3 229
CB719759.1 - 3 289

View Full Product Details