Gene Transcripts

Gene Symbol
Krt23
Entrez Gene ID
287678
Gene Name
keratin 23, type I
Gene Aliases
Ka23, Krt1-23
Chromosome Location
Chr. 10: 87391623 - 87407634 on Build Rnor_6.0
UniGene ID
Rn.137736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008753.1 NP_001008753.1 5, 6 920

View Full Product Details