Gene Transcripts

Gene Symbol
Xbp1
Entrez Gene ID
289754
Gene Name
X-box binding protein 1
Gene Aliases
HTF
Chromosome Location
Chr. 14: 85753736 - 85758820 on Build Rnor_6.0
UniGene ID
Rn.101044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004210.2 NP_001004210.1 3 343
NM_001271731.1 NP_001258660.1 3 343
GenBank mRNA BC079450.1 - 3 324
BU670819.1 - 3 343

View Full Product Details