Gene Transcripts
- Gene Symbol
- Olr1621
- Entrez Gene ID
- 290020
- Gene Name
- olfactory receptor 1621
- Gene Aliases
- Olfr745, ratchr15-26339835-26340800_ORF
- Chromosome Location
- Chr. 15: 27421954 - 27422967 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_223986.7 |
XP_223986.6
|
1 |
428 |
|
View Full Product Details