Gene Transcripts

Gene Symbol
Etfb
Entrez Gene ID
292845
Gene Name
electron-transfer-flavoprotein, beta polypeptide
Gene Aliases
Chromosome Location
Chr. 1: 98472776 - 98486940 on Build Rnor_6.0
UniGene ID
Rn.60152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004220.1 NP_001004220.1 2 115
GenBank mRNA BC079351.1 - 2 115

View Full Product Details