Gene Transcripts

Gene Symbol
RGD1309036
Entrez Gene ID
292874
Gene Name
hypothetical LOC292874
Gene Aliases
Chromosome Location
Chr. 1: 100346096 - 100360635 on Build Rnor_6.0
UniGene ID
Rn.23075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106255.1 NP_001099725.1 5 1003
XM_006228989.2 XP_006229051.1 5 1185
XM_008759332.1 XP_008757554.1 4 802

View Full Product Details