Gene Transcripts

Gene Symbol
Cars
Entrez Gene ID
293638
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
Chromosome Location
Chr. 1: 216759367 - 216801651 on Build Rnor_6.0
UniGene ID
Rn.14865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106319.1 NP_001099789.1 14 1536
XM_006230713.2 XP_006230775.1 15 1792
GenBank mRNA BC166707.1 - 14 1536

View Full Product Details