Gene Transcripts

Gene Symbol
Bcas2
Entrez Gene ID
295334
Gene Name
breast carcinoma amplified sequence 2
Gene Aliases
Chromosome Location
Chr. 2: 205662633 - 205670514 on Build Rnor_6.0
UniGene ID
Rn.1225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106458.2 NP_001099928.1 4 426
GenBank mRNA BC169122.1 - 4 426
FQ226799.1 - 4 418

View Full Product Details