Gene Transcripts

Gene Symbol
Olr773
Entrez Gene ID
296026
Gene Name
olfactory receptor 773
Gene Aliases
ratchr3-97221052-97221990_ORF
Chromosome Location
Chr. 3: 102947730 - 102948668 on Build Rnor_6.0
UniGene ID
Rn.142757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000373.1 NP_001000373.1 1 643

View Full Product Details