Gene Transcripts

Gene Symbol
Vps28
Entrez Gene ID
300052
Gene Name
vacuolar protein sorting 28 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 7: 117684339 - 117688187 on Build Rnor_6.0
UniGene ID
Rn.36912
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130492.1 NP_001123964.1 3, 4 229
GenBank mRNA BC168742.1 - 3, 4 175
FQ225519.1 - 3, 4 196

View Full Product Details