Gene Transcripts

Gene Symbol
Mrpl3
Entrez Gene ID
300974
Gene Name
mitochondrial ribosomal protein L3
Gene Aliases
Chromosome Location
Chr. 8: 113603533 - 113626893 on Build Rnor_6.0
UniGene ID
Rn.6950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106852.1 NP_001100322.1 5 738
GenBank mRNA FQ233139.1 - 1 92

View Full Product Details