Gene Transcripts

Gene Symbol
Copg2
Entrez Gene ID
301742
Gene Name
coatomer protein complex, subunit gamma 2
Gene Aliases
RGD1566215
Chromosome Location
Chr. 4: 58064394 - 58195242 on Build Rnor_6.0
UniGene ID
Rn.216502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106929.2 NP_001100399.1 2, 3 197
GenBank mRNA FM104860.1 - 2, 3 133
FN805881.1 - 2, 3 117

View Full Product Details